Order Entry
Export
ContactUsLinkComponent
Anti-IgG Goat Polyclonal Antibody (APC (Allophycocyanin))
Anti-IgG Goat Polyclonal Antibody (APC (Allophycocyanin))
Certificates
Anti-IgG Goat Polyclonal Antibody (APC (Allophycocyanin))

Specifications

  • Catalog No:
  • COBSD3-112-1MG
  • COBSD3-112-100
  • Antigen symbol:
  • IgG
  • IgG
  • Antibody type:
  • Secondary
  • Secondary
  • Clonality:
  • Polyclonal
  • Polyclonal
  • Conjugation:
  • APC (Allophycocyanin)
  • APC (Allophycocyanin)
  • Reactivity:
  • Mouse
  • Mouse
  • Host:
  • Goat
  • Goat
  • Isotype:
  • IgG
  • IgG
  • ELISA:
  • Yes
  • Yes
  • Flow cytometry:
  • Yes
  • Yes
  • Cross adsorption:
  • Bovine, Rabbit
  • Bovine, Rabbit
  • Storage temperature:
  • Cool
  • Cool
  • Shipping temperature:
  • Ice
  • Ice

Specifications

Related Information