Anti-DYKDDDDK Tag Mouse Monoclonal Antibody (Europium 1024) [clone: M2]
Supplier: Columbia Biosciences
Certificates
About this item
Type: Tag
Antigen: DYKDDDDK Tag (DYKDDDDK tag)
Clonality: monoclonal
Clone: M2
Conjugation: Europium 1024
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity:
Specifications
- Catalog No:
- COBSD17-1718-20
- COBSD17-1718-1MG
- COBSD17-1718-50
- Antigen symbol:
- DYKDDDDK
- DYKDDDDK
- DYKDDDDK
- Antigen name:
- DYKDDDDK tag
- DYKDDDDK tag
- DYKDDDDK tag
- Antibody type:
- Tag
- Tag
- Tag
- Clonality:
- Monoclonal
- Monoclonal
- Monoclonal
- Conjugation:
- Europium 1024
- Europium 1024
- Europium 1024
- Clone:
- M2
- M2
- M2
- Host:
- Mouse
- Mouse
- Mouse
- Isotype:
- IgG1 kappa
- IgG1 kappa
- IgG1 kappa
- Cross adsorption:
- none
- none
- none
- Storage temperature:
- Cool
- Cool
- Cool
- Shipping temperature:
- Ice
- Ice
- Ice