Anti-S Tag Rabbit Polyclonal Antibody (DyLight® 650)
COBSD10-1831
- Antibody type:Tag
- Antigen name:S tag
- Antigen symbol:S
- Clonality:Polyclonal
- Conjugation:DyLight® 650
- ELISA:Yes
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Western blot:Yes
- Cross adsorption:none
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µG
Type: Tag
Antigen: S Tag (S tag)
Clonality: polyclonal
Clone:
Conjugation: DyLight® 650
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: