Order Entry
Export
ContactUsLinkComponent
Anti-Alpha-1-Antichymotrypsin Sheep Polyclonal Antibody
Anti-Alpha-1-Antichymotrypsin Sheep Polyclonal Antibody
Catalog # IMMRIR2692
Supplier:  ImmunoReagents
Anti-Alpha-1-Antichymotrypsin Sheep Polyclonal Antibody
Catalog # IMMRIR2692
Supplier:  ImmunoReagents

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Alpha-1-Antichymotrypsin
  • Antigen symbol:
    ACT
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Flow cytometry:
    Yes
  • Host:
    Sheep
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Format:
    Purified IgG
  • Form:
    Clear, colorless liquid, 0.2 µm filtered
  • Antigen synonyms:
    MGC88254|GIG24|GIG25
  • Storage buffer:
    10 mM Sodium Phosphate, 0.15 M Sodium Chloride, pH 7.2
  • Storage temperature:
    2-8 °C
  • Concentration:
    > 4,5 mg/ml (E 1% at 280 nm = 13,0)
  • Shipping temperature:
    2-8 °C
  • Immunogen:
    Human Alpha-1-Antichymotrypsin
  • Tested applications:
    Fluorescence-Activated Cell Sorting, Immunodiffusion, Labeling
  • Purification:
    Affinity purified using solid phase Human Alpha-1-Antichymotrypsin
  • Size:
    1,0 mg
  • Pk:
    1 mg

Specifications

About this item

Type: Primary
Antigen: Alpha-1-Antichymotrypsin
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: Human