Specifications
- Antibody type:Secondary
- Antigen name:Immunoglobulin G
- Antigen symbol:IgG
- Clonality:Polyclonal
- Conjugation:DyLight® 550
- ELISA:Yes
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Mouse
- Western blot:Yes
- Cross adsorption:Yes
- Format:Purified IgG
- Form:Lyophilized
- Storage buffer:10 mM Sodium phosphate, 0,15 M Sodium chloride, pH 7,2, 1 % (w/v) BSA, Protease/IgG free
- Storage temperature:2 - 8 °C
- Concentration:1,0 mg/ml (E 1% at 280 nm = 13,0)
- Shipping temperature:2 - 8 °C
- Immunogen:Mouse IgG
- Tested applications:Fluoresence ELISA, Fluorescence-Activated Cell Sorting
- Purification:Affinity purified using solid phaseMouse IgG
- Size:1,0 mg
- Pk:1 mg
Specifications
About this item
The ImmunoReagents' DyLight® 550 conjugates provide vibrant orange-to-red fluorescence with better performance than other rhodamine derivatives, including Alexa Fluor 555, TRITC and Cy3 for fluorescent applications. DyLight® 550 conjugates have tight excitation/emission spectra that are ideal for multicolor applications. DyLight® 550 conjugated secondary antibodies are used in fluorescence microscopy, flow cytometry, Western blotting, ELISA, high-content screening and other array platforms. Excitation/Emission = 562nm / 576nm Emission Color = Orange to Red (Similar Dyes: Alexa Fluor 555, Cy3, TRITC)
- Origin: US
ImmunoReagents' antibodies are suitable for most immunoassay formats and purified, cross adsorbed and conjugated based on the final product specfications.
Products will be accompanied with a standard product CoA to include application, purity and cross reactivity profiles.
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: DyLight® 550
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse