Anti-IgG Rabbit Polyclonal Antibody
Product Details & Documents
ImmunoReagents is a manufacturer of quality research and production reagents for the Life Science Industries. Product lines include a wide range of antibodies used in ELISA, immunohistochemistry (IHC), western blotting, flow cytometry (FACS), immunofluorescence and microscopy. Antibodies produced cover a broad range of Immunoglobulins from various species cross absorbed to provide high specificity.
- Origin: US
Products will be accompanied with a standard product CoA to include application, purity and cross reactivity profiles.
ImmunoReagents' antibodies are suitable for most immunoassay formats and purified, cross adsorbed and conjugated based on the final product specfications.
Certifications: (EC) 142/2011 approved by APHIS, Reference Number is NC-TEC-0002
Ordering information: Products can by customised based on specific assay requirements: contact your VWR representative for more information.
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Hamster
Documents
RbxHa-003-E
Datasheet
Specifications
- Size:1,0 mg
- Western blot:Yes
- Clonality:Polyclonal
- Flow cytometry:Yes
- ImmunoPrecipitation:Yes
- Form:Clear, colorless liquid, 0.2 µm filtered
- ImmunoFluorescence:Yes
- Storage temperature:2...8 °C
- Antigen name:Immunoglobulin G
- Format:Purified IgG
- Immunogen:Hamster IgG
- Tested applications:Fluorescence-Activated Cell Sorting, Immunodiffusion, Labeling
- Storage buffer:10 mM Sodium Phosphate, 0,15 M Sodium Chloride, pH 7,2
- Purification:Affinity purified using solid phaseHamster IgG
- Antibody type:Secondary
- Isotype:IgG
- Host:Rabbit
- Concentration:> 4,5 mg/ml (E 1% at 280 nm = 14,0)
- Cross adsorption:Min x human serum proteins
- Shipping temperature:2-8 °C
- Cat. No.:IMMRIR2559
- Reactivity:Hamster
- ImmunoChemistry:Yes
- Conjugation:Unconjugated
- Antigen symbol:IgG
- ELISA:Yes



