Specifications
- Antibody type:Secondary
- Antigen name:Immunoglobulin G
- Antigen symbol:IgG
- Clonality:Polyclonal
- Conjugation:AP (Alkaline Phosphatase)
- ELISA:Yes
- Flow cytometry:Yes
- Gel shift:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Hamster
- Western blot:Yes
- Cross adsorption:No
- Format:Purified IgG
- Form:Clear, colorless liquid
- Storage buffer:30 mM Triethanolamine, pH 7,2, 5 mM Magnesium Chloride, 0,1 mM Zinc chloride, 1 % (w/v) BSA, Protease/IgG free
- Storage temperature:2 - 8 °C
- Concentration:1,50 mg/ml (E 1% at 280 nm = 13,0)
- Shipping temperature:2 - 8 °C
- Immunogen:Hamster IgG
- Purification:Affinity purified using solid phaseHamster IgG
- Size:1,0 mg
- Pk:1 mg
Specifications
About this item
ImmunoReagents’ Alkaline Phosphatase (AP) antibody conjugates are used in ELISA, Immunohistochemistry (IHC) and Western blotting techniques. AP is a 140 kDa enzyme that changes color in the presence of PNPP or BCIP/NBT substrates. These conjugates have a linear reaction rate thus allowing the researcher to increase sensitivity by prolonging substrate incubation times.
- Origin: US
ImmunoReagents' antibodies are suitable for most immunoassay formats and purified, cross adsorbed and conjugated based on the final product specfications.
Products will be accompanied with a standard product CoA to include application, purity and cross reactivity profiles.
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Hamster