Order Entry
Export
ContactUsLinkComponent
Anti-Cardiac Myoglobin Goat Polyclonal Antibody
Anti-Cardiac Myoglobin Goat Polyclonal Antibody
Catalog # IMMRIR2108
Supplier:  ImmunoReagents
Anti-Cardiac Myoglobin Goat Polyclonal Antibody
Catalog # IMMRIR2108
Supplier:  ImmunoReagents

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Cardiac Myoglobin
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Flow cytometry:
    Yes
  • Host:
    Goat
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Format:
    Purified IgG
  • Form:
    Clear, colourless liquid, 0,2 µm filtered
  • Antigen synonyms:
    Purified Human Cardiac Myoglobin
  • Storage buffer:
    10 mM sodium phosphate, 0,15 M sodium chloride, pH 7,2
  • Storage temperature:
    2...8 °C
  • Concentration:
    >10 mg/ml (E 1% at 280 nm = 1,3)
  • Shipping temperature:
    2...8 °C
  • Immunogen:
    Human Cardiac Myoglobin
  • Tested applications:
    Fluorescence-Activated Cell Sorting, Immunodiffusion, Labeling
  • Purification:
    Proprietary 2-step fractionation procedure
  • Size:
    1,0 ml
  • Pk:
    1 ml

Specifications

About this item

ImmunoReagents is a manufacturer of quality research and production reagents for the Life Science Industries. Product lines include a wide range of antibodies used in ELISA, immunohistochemistry (IHC), western blotting, flow cytometry (FACS), immunofluorescence and microscopy.  Antibodies produced cover a broad range of Immunoglobulins from various species cross absorbed to provide high specificity.  

  • Origin: US

Products will be accompanied with a standard product CoA to include application, purity and cross reactivity profiles.

ImmunoReagents' antibodies are suitable for most immunoassay formats and purified, cross adsorbed and conjugated based on the final product specfications.

Certifications: (EC) 142/2011 approved by APHIS, Reference Number is NC-TEC-0002
Ordering information: Products can by customised based on specific assay requirements: contact your VWR representative for more information.

Type: Primary
Antigen: Cardiac Myoglobin
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: