Order Entry
Export
Orders LinkContactUsLinkComponent
Anti-IgG/IgM Goat Polyclonal Antibody

Anti-IgG/IgM Goat Polyclonal Antibody

1,0 mg
Catalog # IMMRIR2032
Supplier:   ImmunoReagents
Anti-IgG/IgM Goat Polyclonal Antibody
Catalog # IMMRIR2032
Supplier:  ImmunoReagents
Anti-IgG/IgM Goat Polyclonal Antibody
Anti-IgG/IgM Goat Polyclonal Antibody

1,0 mg

Price
153,00 €
Log in for availability
1 * 1 mg
Product Details & Documents

ImmunoReagents is a manufacturer of quality research and production reagents for the Life Science Industries. Product lines include a wide range of antibodies used in ELISA, immunohistochemistry (IHC), western blotting, flow cytometry (FACS), immunofluorescence and microscopy.  Antibodies produced cover a broad range of Immunoglobulins from various species cross absorbed to provide high specificity.  

  • Origin: US

Products will be accompanied with a standard product CoA to include application, purity and cross reactivity profiles.

ImmunoReagents' antibodies are suitable for most immunoassay formats and purified, cross adsorbed and conjugated based on the final product specfications.

Certifications: (EC) 142/2011 approved by APHIS, Reference Number is NC-TEC-0002

Ordering information: Products can by customised based on specific assay requirements: contact your VWR representative for more information.

Type: Secondary
Antigen: IgG/IgM
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG/IgM
Reactivity: Mouse

Documents

Specifications
  • Size:
    1,0 mg
  • Western blot:
    Yes
  • Clonality:
    Polyclonal
  • Flow cytometry:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Form:
    Clear, colourless liquid, 0,2 µm filtered
  • ImmunoFluorescence:
    Yes
  • Storage temperature:
    2...8 °C
  • Antigen name:
    Immunoglobulin G And M
  • Format:
    Purified IgG
  • Immunogen:
    Mouse IgG + IgM
  • Tested applications:
    Fluorescence-Activated Cell Sorting, Immunodiffusion, Labeling
  • Storage buffer:
    10 mM sodium phosphate, 0,15 M sodium chloride, pH 7,2
  • Purification:
    Affinity purified using solid phaseMouse IgG + IgM
  • Antibody type:
    Secondary
  • Isotype:
    IgG
  • Host:
    Goat
  • Concentration:
    >4,5 mg/ml (E 1% at 280 nm = 13,0)
  • Cross adsorption:
    Min x w/human IgG or serum proteins
  • Shipping temperature:
    2...8 °C
  • Cat. No.:
    IMMRIR2032
  • Reactivity:
    Mouse
  • ImmunoChemistry:
    Yes
  • Conjugation:
    Unconjugated
  • Antigen symbol:
    IgG + IgM
  • ELISA:
    Yes