Order Entry
Export
ContactUsLinkComponent
Anti-IgM µ Goat Polyclonal Antibody (AP (Alkaline Phosphatase))
Anti-IgM µ Goat Polyclonal Antibody (AP (Alkaline Phosphatase))
Catalog # IMMRIR1771
Supplier:  ImmunoReagents
Anti-IgM µ Goat Polyclonal Antibody (AP (Alkaline Phosphatase))
Catalog # IMMRIR1771
Supplier:  ImmunoReagents

Specifications

  • Antibody type:
    Secondary
  • Antigen name:
    Immunoglobulin M mu-chain
  • Antigen symbol:
    IgM
  • Clonality:
    Polyclonal
  • Conjugation:
    AP (Alkaline Phosphatase)
  • ELISA:
    Yes
  • Flow cytometry:
    Yes
  • Gel shift:
    Yes
  • Host:
    Goat
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Epitope:
    µ-chain
  • Cross adsorption:
    Yes
  • Format:
    Purified IgG
  • Form:
    Clear, colorless liquid
  • Antibody fragment:
    F(ab')2
  • Storage buffer:
    30 mM Triethanolamine, pH 7,2, 5 mM Magnesium Chloride, 0,1 mM Zinc Chloride, 1 % (w/v) BSA, Protease/IgG free
  • Storage temperature:
    2…8 °C
  • Concentration:
    1,50 mg/ml (E 1% at 280 nm = 13,0)
  • Shipping temperature:
    2…8 °C
  • Immunogen:
    Human IgM, µ chain
  • Purification:
    Affinity purified using solid phase Human IgM
  • Size:
    0,5 mg
  • Pk:
    0,5 mg

Specifications

About this item

ImmunoReagents Alkaline Phosphatase (AP) antibody conjugates are used in ELISA, Immunohistochemistry (IHC) and Western blotting techniques. AP is a 140 kDa enzyme that changes color in the presence of PNPP or BCIP/NBT substrates. These conjugates have a linear reaction rate thus allowing the researcher to increase sensitivity by prolonging substrate incubation times.

  • Origin: US

ImmunoReagents antibodies are suitable for most immunoassay formats and purified, cross adsorbed and conjugated based on the final product specfications.

Products will be accompanied with a standard product CoA to include application, purity and cross reactivity profiles.

Certifications: (EC) 142/2011 approved by APHIS, Reference Number is NC-TEC-0002.
Ordering information: Products can by customized based on specific assay requirements: contact your VWR representative for more information.

Type: Secondary
Antigen: IgM µ
Clonality: Polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope:
Host: Goat
Isotype: IgM
Reactivity: Human