Order Entry
Export
ContactUsLinkComponent
Anti-IgA/IgG/IgM Goat Polyclonal Antibody
Anti-IgA/IgG/IgM Goat Polyclonal Antibody
Catalog # IMMRIR1747
Supplier:  ImmunoReagents
undefined
Anti-IgA/IgG/IgM Goat Polyclonal Antibody
Catalog # IMMRIR1747
Supplier:  ImmunoReagents

Specifications

  • Antibody type:
    Secondary
  • Antigen name:
    Immunoglobulin A, G And M
  • Antigen symbol:
    IgG+IgA+IgM
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Flow cytometry:
    Yes
  • Host:
    Goat
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Format:
    Purified IgG
  • Form:
    Clear, colorless liquid, 0,2 µm filtered
  • Antibody fragment:
    F(ab')2
  • Storage buffer:
    10 mM Sodium Phosphate, 0,15 M Sodium Chloride, pH 7,2
  • Storage temperature:
    2…8 °C
  • Concentration:
    >4,5 mg/ml (E 1% at 280 nm = 13,0)
  • Shipping temperature:
    2…8 °C
  • Immunogen:
    Human IgG + IgA + IgM
  • Tested applications:
    Fluorescence-Activated Cell Sorting, Immunodiffusion, Labeling
  • Purification:
    Affinity purified using solid phase Human IgA + IgG + IgM
  • Size:
    1,0 mg
  • Pk:
    2 mg

Specifications

About this item

ImmunoReagents is a manufacturer of quality research and production reagents for the Life Science Industries. Product lines include a wide range of antibodies used in ELISA, immunohistochemistry (IHC), western blotting, flow cytometry (FACS), immunofluorescence and microscopy. Antibodies produced cover a broad range of Immunoglobulins from various species cross absorbed to provide high specificity.

  • Origin: US

Products will be accompanied with a standard product CoA to include application, purity and cross reactivity profiles.

ImmunoReagents antibodies are suitable for most immunoassay formats and purified, cross adsorbed and conjugated based on the final product specfications.

Certifications: (EC) 142/2011 approved by APHIS, Reference Number is NC-TEC-0002.
Ordering information: Products can by customized based on specific assay requirements: contact your VWR representative for more information.

Type: Secondary
Antigen: IgG/IgA/IgM
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG/IgA/IgM
Reactivity: Human