Anti-IgG Donkey Polyclonal Antibody
ImmunoReagents is a manufacturer of quality research and production reagents for the Life Science Industries. Product lines include a wide range of antibodies used in ELISA, immunohistochemistry (IHC), western blotting, flow cytometry (FACS), immunofluorescence and microscopy. Antibodies produced cover a broad range of Immunoglobulins from various species cross absorbed to provide high specificity.
- Origin: US
Products will be accompanied with a standard product CoA to include application, purity and cross reactivity profiles.
ImmunoReagents' antibodies are suitable for most immunoassay formats and purified, cross adsorbed and conjugated based on the final product specfications.
: (EC) 142/2011 approved by APHIS, Reference Number is NC-TEC-0002.
: Products can by customized based on specific assay requirements: contact your VWR representative for more information.
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Donkey
Isotype: IgG
Reactivity: Sheep
- Size:2,0 mg
- Western blot:Yes
- Clonality:Polyclonal
- Flow cytometry:Yes
- ImmunoPrecipitation:Yes
- Form:Clear, colorless liquid, 0,2 µm filtered
- ImmunoFluorescence:Yes
- Storage temperature:2...8 °C
- Antigen name:Immunoglobulin G
- Format:Purified IgG
- Immunogen:Sheep IgG
- Tested applications:Fluorescence-Activated Cell Sorting, Immunodiffusion, Labeling
- Storage buffer:10 mM Sodium Phosphate, 0,15 M Sodium Chloride, pH 7,2
- Purification:Affinity purified using solid phaseSheep IgG
- Antibody type:Secondary
- Isotype:IgG
- Host:Donkey
- Concentration:>4,5 mg/ml (E 1% at 280 nm = 14,0)
- Cross adsorption:Yes
- Shipping temperature:2...8 °C
- Cat. No.:IMMRIR1298
- Reactivity:Sheep
- ImmunoChemistry:Yes
- Conjugation:Unconjugated
- Antigen symbol:IgG
- ELISA:Yes



