Order Entry
Export
ContactUsLinkComponent
 
Anti-IgG Donkey Polyclonal Antibody (AP (Alkaline Phosphatase))
  IMMRIR1292
undefined
Anti-IgG Donkey Polyclonal Antibody (AP (Alkaline Phosphatase))
  IMMRIR1292

 

  • Antibody type:
    Secondary
  • Antigen name:
    Immunoglobulin G
  • Antigen symbol:
    IgG
  • Clonality:
    Polyclonal
  • Conjugation:
    AP (Alkaline Phosphatase)
  • ELISA:
    Yes
  • Flow cytometry:
    Yes
  • Gel shift:
    Yes
  • Host:
    Donkey
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Sheep
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Format:
    Purified IgG
  • Form:
    Clear, colorless liquid
  • Storage buffer:
    30 mM Triethanolamine, pH 7.2, 5 mM Magnesium Chloride, 0.1 mM Zinc Chloride, 1 % (w/v) BSA, Protease/IgG free
  • Storage temperature:
    2-8 °C
  • Concentration:
    1,50 mg/ml (E 1% at 280 nm = 13,0)
  • Shipping temperature:
    2-8 °C
  • Immunogen:
    Sheep IgG
  • Purification:
    Affinity purified using solid phaseSheep IgG
  • Size:
    1,0 mg
  • Pk:
    1 mg

 

 

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope:
Host: Donkey
Isotype: IgG
Reactivity: Sheep