Specifications
- Antibody type:Secondary
- Antigen name:Immunoglobulin G
- Antigen symbol:IgG
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- Host:Donkey
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Rabbit
- Western blot:Yes
- Cross adsorption:Yes
- Format:Purified IgG
- Form:Clear, colorless liquid, 0,2 µm filtered
- Storage buffer:10 mM Sodium Phosphate, 0,15 M Sodium Chloride, pH 7,2
- Storage temperature:2...8 °C
- Concentration:>4,5 mg/ml (E 1% at 280 nm = 13,0)
- Shipping temperature:2...8 °C
- Immunogen:Rabbit IgG
- Tested applications:Fluorescence-Activated Cell Sorting, Immunodiffusion, Labeling
- Purification:Affinity purified using solid phaseRabbit IgG
- Size:2,0 mg
- Pk:2 mg
Specifications
About this item
ImmunoReagents is a manufacturer of quality research and production reagents for the Life Science Industries. Product lines include a wide range of antibodies used in ELISA, immunohistochemistry (IHC), western blotting, flow cytometry (FACS), immunofluorescence and microscopy. Antibodies produced cover a broad range of Immunoglobulins from various species cross absorbed to provide high specificity.
- Origin: US
Products will be accompanied with a standard product CoA to include application, purity and cross reactivity profiles.
ImmunoReagents' antibodies are suitable for most immunoassay formats and purified, cross adsorbed and conjugated based on the final product specfications.
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Donkey
Isotype: IgG
Reactivity: Rabbit