Order Entry
Export
ContactUsLinkComponent
Anti-TUBA1A Mouse Monoclonal Antibody [clone: TU-16]
Anti-TUBA1A Mouse Monoclonal Antibody [clone: TU-16]
Catalog # USBIT9154-06M
Supplier:  US Biological
Anti-TUBA1A Mouse Monoclonal Antibody [clone: TU-16]
Catalog # USBIT9154-06M
Supplier:  US Biological

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    tubulin, alpha 1a
  • Antigen symbol:
    TUBA1A
  • Clonality:
    Monoclonal
  • Clone:
    TU-16
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgM
  • Reactivity:
    Pig
  • Western blot:
    Yes
  • Form:
    liquid
  • Storage buffer:
    PBS, 15mM sodium azide, pH 7,4.
  • Storage temperature:
    Cool
  • Concentration:
    ~1 mg/ml
  • Shipping temperature:
    Ice
  • Purification:
    purified by ammonium sulfate precipitation
  • Size:
    50 µg
  • Pk:
    50 µG

Specifications

About this item

Recommended dilution: Western Blot: 1-2?g/ml; Immunochemistry: 10 ?g/ml; Immunoprecipitation: 1-2?g/ml

Type: Primary
Antigen: TUBA1A (tubulin, alpha 1a)
Clonality: monoclonal
Clone: TU-16
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: Pig