Anti-TNNT3 Mouse Monoclonal Antibody [clone: T1/61]
Catalog # USBIT8665-30J
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:troponin T type 3 (skeletal, fast)
- Antigen symbol:TNNT3
- Clonality:Monoclonal
- Clone:T1/61
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Chicken
- Western blot:Yes
- Form:liquid
- Storage buffer:PBS, 0,09% sodium azide.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein G chromatography
- Size:50 µg
- Pk:50 µG
Specifications
About this item
Recommended dilution: Immunochemistry: 10 ?g/ml
Type: Primary
Antigen: TNNT3 (troponin T type 3 (skeletal, fast))
Clonality: monoclonal
Clone: T1/61
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Chicken