Anti-NTRK3 Goat Polyclonal Antibody (PE (Phycoerythrin))
USBIT8663-26
- Antibody type:Primary
- Antigen name:neurotrophic tyrosine kinase, receptor, type 3
- Antigen symbol:NTRK3
- Clonality:Polyclonal
- Conjugation:PE (Phycoerythrin)
- Flow cytometry:Yes
- Host:Goat
- Isotype:IgG
- Reactivity:Human
- Form:liquid
- Storage buffer:PBS, 0,01% sodium azide.
- Storage temperature:Cool
- Concentration:~0,05 mg/ml
- Shipping temperature:Ice
- Purification:purified
- Size:100 Tests
- Pk:100 Tests
Do not freeze!
Type: Primary
Antigen: NTRK3 (neurotrophic tyrosine kinase, receptor, type 3)
Clonality: polyclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human