Anti-TRA1-60 Mouse Monoclonal Antibody [clone: 5C2]
Catalog # USBIT8115
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen symbol:TRA1-60
- Clonality:Monoclonal
- Clone:5C2
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgM
- Reactivity:Human
- Western blot:Yes
- Form:liquid
- Storage buffer:PBS, pH 8.0, 0,05% sodium azide.
- Storage temperature:Cool
- Concentration:~1 mg/ml
- Shipping temperature:Ice
- Purification:purified
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Recommended dilution: Immunofluorescence: 10-20 ?g/ml
Type: Primary
Antigen: TRA1-60
Clonality: monoclonal
Clone: 5C2
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: Human