Anti-Tissue Factor Pathway Inhibitor Sheep Polyclonal Antibody
USBIT5576-05-10
- Antibody type:Primary
- Antigen name:Tissue Factor Pathway Inhibitor
- Antigen symbol:TFPI
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Sheep
- Isotype:IgG
- Reactivity:Human
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:10 mg
- Pk:10 mg
Type: Primary
Antigen: Tissue Factor Pathway Inhibitor
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: Human