Anti-TID 1 Mouse Monoclonal Antibody [clone: 4i17]
USBIT5498-55A
- Antibody type:Primary
- Antigen symbol:TID 1
- Clonality:Monoclonal
- Clone:4i17
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoPrecipitation:Yes
- Isotype:IgM
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Format:ascites
- Form:liquid
- Storage buffer:10mMsodium HEPES, pH 7,5, 150 mM NaCl, 100ug/ml BSA and 50% glycerol.
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µl
- Pk:100 µl
Recommended dilution: Western Blot: 1:1000; Immunoprecipitation: 1:100
Type: Primary
Antigen: TID 1
Clonality: monoclonal
Clone: 4i17
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: Human, Mouse, Rat