Order Entry
Export
ContactUsLinkComponent
Anti-TPO Mouse Monoclonal Antibody [clone: 1.B.10]
Anti-TPO Mouse Monoclonal Antibody [clone: 1.B.10]
Catalog # USBIT5399-01-250
Supplier:  US Biological
Anti-TPO Mouse Monoclonal Antibody [clone: 1.B.10]
Catalog # USBIT5399-01-250
Supplier:  US Biological

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Thyroid Peroxidase
  • Antigen symbol:
    TPO
  • Clonality:
    Monoclonal
  • Clone:
    1.B.10
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG1
  • Reactivity:
    Human
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    250 µg
  • Pk:
    250 µG

Specifications

About this item

Type: Primary
Antigen: TPO (thyroid peroxidase)
Clonality: monoclonal
Clone: 1.B.10
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human