Order Entry
Export
ContactUsLinkComponent
Anti-T Cells, Pan Mouse Monoclonal Antibody [clone: 15-16A1]
Anti-T Cells, Pan Mouse Monoclonal Antibody [clone: 15-16A1]
Catalog # USBIT2010-31A
Supplier:  US Biological
Anti-T Cells, Pan Mouse Monoclonal Antibody [clone: 15-16A1]
Catalog # USBIT2010-31A
Supplier:  US Biological

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    T Cells, Pan
  • Clonality:
    Monoclonal
  • Clone:
    15-16A1
  • Conjugation:
    Unconjugated
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG1
  • Reactivity:
    Rat
  • Form:
    liquid
  • Storage temperature:
    Cool
  • Concentration:
    ~0,1 mg/ml
  • Shipping temperature:
    Ice
  • Purification:
    purified by Protein G chromatography
  • Size:
    1 ml
  • Pk:
    1 mL

Specifications

About this item

Recommended dilution: Flow Cytometry: 1:10; Immunochemistry: 1:100

Type: Primary
Antigen: T Cells, Pan
Clonality: monoclonal
Clone: 15-16A1
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Rat