Anti-TAR DNA Binding Protein 43 Rat Monoclonal Antibody [clone: 10H31]
Catalog # USBIT1019-47C
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:TAR DNA-binding protein 43
- Antigen symbol:TDP43
- Clonality:Monoclonal
- Clone:10H31
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rat
- Isotype:IgG2a kappa
- Reactivity:Human
- Western blot:Yes
- Form:liquid
- Storage buffer:0,1M Tris-glycine, pH 7,4, 0,15M sodium chloride, 0,05% sodium azide.
- Storage temperature:Cool
- Concentration:~0,5 mg/ml
- Shipping temperature:Ice
- Purification:purified by Protein G chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Recommended dilution: Western Blot: 1:1000
Type: Primary
Antigen: TAR DNA Binding Protein 43
Clonality: monoclonal
Clone: 10H31
Conjugation: unconjugated
Epitope:
Host: Rat
Isotype: IgG2a Kappa
Reactivity: Human