Anti-TAR DNA-binding Protein-43 Mouse Monoclonal Antibody [clone: K1B8]
Catalog # USBIT1019-41B
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:TAR DNA-binding protein 43
- Antigen symbol:TDP43
- Clonality:Monoclonal
- Clone:K1B8
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Human
- Western blot:Yes
- Form:liquid
- Amino acid number:1 to 260
- Storage buffer:PBS, pH 7,4, 0,1% sodium azide.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein G chromatography
- Size:50 µg
- Pk:50 µG
Specifications
About this item
Recommended dilution: Western Blot:1:1000-1:2000; Immunochemistry:10 ?g/ml
Type: Primary
Antigen: TAR DNA-binding Protein-43
Clonality: monoclonal
Clone: K1B8
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human