Anti-TAR DNA-binding Protein-43 Rabbit Polyclonal Antibody
Catalog # USBIT1019-41A
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:TAR DNA-binding protein 43
- Antigen symbol:TDP43
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Form:liquid
- Storage buffer:PBS, 0,02% sodium azide.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by immunoaffinity chromatography
- Size:50 µg
- Pk:50 µG
Specifications
About this item
Recommended dilution: Western Blot: 1 ?g/ml; Immunochemistry: 5 ?g/ml
Type: Primary
Antigen: TAR DNA-binding Protein-43
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human