Anti-Tacrolimus 32 Sheep Polyclonal Antibody
USBIT0501-25
- Antibody type:Primary
- Antigen symbol:Tacrolimus 32
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Sheep
- Isotype:IgG
- Form:liquid
- Storage buffer:PBS, pH 7,2, 0,09% sodium azide.
- Storage temperature:Cool
- Concentration:~1 mg/ml
- Shipping temperature:Ice
- Purification:purified
- Size:100 µl
- Pk:100 µl
Recommended dilution: ELISA: 0. 625 ?g/ml
Type: Primary
Antigen: Tacrolimus 32
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: