Order Entry
Export
ContactUsLinkComponent
Anti-Streptavidin Mouse Monoclonal Antibody [clone: Strep-10]
Anti-Streptavidin Mouse Monoclonal Antibody [clone: Strep-10]
Catalog # USBIS7973-71C
Supplier:  US Biological
Anti-Streptavidin Mouse Monoclonal Antibody [clone: Strep-10]
Catalog # USBIS7973-71C
Supplier:  US Biological

Specifications

  • Antibody type:
    Tag
  • Antigen symbol:
    Streptavidin
  • Clonality:
    Monoclonal
  • Clone:
    Strep-10
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG1
  • Format:
    Serum
  • Form:
    liquid
  • Storage buffer:
    1% BSA, 0,1% sodium azide.
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    1 ml
  • Pk:
    1 mL

Specifications

About this item

Recommended dilution: Immunochemistry: 1:20

Type: Tag
Antigen: Streptavidin
Clonality: Monoclonal
Clone: Strep-10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: