Anti-STAT 1 Mouse Monoclonal Antibody [clone: 5J111]
Catalog # USBIS7969-05G
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen symbol:STAT 1
- Clonality:Monoclonal
- Clone:5J111
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:liquid
- Storage buffer:PBS1% BSA, pH 7,2, 0,1% sodium azide.
- Storage temperature:Cool
- Concentration:~0,5 mg/ml
- Shipping temperature:Ice
- Purification:purified by affinity chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Recommended dilution: Western Blot: 1 ?g/ml
Type: Primary
Antigen: STAT 1
Clonality: Monoclonal
Clone: 5J111
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human, Mouse, Rat