Order Entry
Export
ContactUsLinkComponent
Anti-SC5b-9 Mouse Monoclonal Antibody [clone: 10F425]
Anti-SC5b-9 Mouse Monoclonal Antibody [clone: 10F425]
Catalog # USBIS7850-26
Supplier:  US Biological
Anti-SC5b-9 Mouse Monoclonal Antibody [clone: 10F425]
Catalog # USBIS7850-26
Supplier:  US Biological

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    SC5b-9
  • Clonality:
    Monoclonal
  • Clone:
    10F425
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG2b
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Form:
    liquid
  • Storage buffer:
    borate buffered saline, 0,1% sodium azide.
  • Storage temperature:
    Cool
  • Concentration:
    ~1 mg/ml
  • Shipping temperature:
    Ice
  • Purification:
    purified by Protein A chromatography
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Type: Primary
Antigen: SC5b-9
Clonality: Monoclonal
Clone: 10F425
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human