Order Entry
Export
ContactUsLinkComponent
Anti-SSA1/Ro-52 Mouse Monoclonal Antibody [clone: 11n177]
Anti-SSA1/Ro-52 Mouse Monoclonal Antibody [clone: 11n177]
  USBIS6600-65C1
Anti-SSA1/Ro-52 Mouse Monoclonal Antibody [clone: 11n177]
  USBIS6600-65C1

 

  • Antibody type:
    Primary
  • Antigen symbol:
    SSA1
  • Clonality:
    Monoclonal
  • Clone:
    11n177
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • Isotype:
    IgG1
  • Reactivity:
    Human
  • Form:
    lyophilized powder
  • Antigen synonyms:
    Ro-52
  • Amino acid number:
    195 to 293
  • Storage buffer:
    PBS, pH 7,4, 5% trehalose.
  • Storage temperature:
    Cool
  • Concentration:
    ~0,5 mg/ml (after reconstitution)
  • Shipping temperature:
    Ice
  • Purification:
    purified by Protein G chromatography
  • Size:
    100 µg
  • Pk:
    100 µG

 

 

Recommended dilution: Immunochemistry: 8-25 ?g/ml

Type: Primary
Antigen: SSA1/Ro-52
Clonality: Monoclonal
Clone: 11n177
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human