Anti-SLC5A6 Rabbit Polyclonal Antibody
USBIS5328-96A
- Antibody type:Primary
- Antigen name:solute carrier family 5 (sodium/multivitamin and iodide cotransporter), member 6
- Antigen symbol:Solute carrier family 5, member 6
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:
- Reactivity:Human
- Environmentally Preferable:
- Form:liquid
- Storage buffer:PBS, 0,1% sodium azide.
- Storage temperature:Cool
- Concentration:~0,5 mg/ml
- Shipping temperature:Ice
- Purification:purified by immunoaffinity chromatography
- Size:50 µg
- Pk:50 µG
Recommended dilution: Immunochemistry: 10-15 ?g/ml
Type: Primary
Antigen: SLC5A6 (solute carrier family 5 (sodium/multivitamin and iodide cotransporter), member 6)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human