Anti-Small Cell Lung Carcinoma, Neural and Endocrine Antigen Mouse Monoclonal Antibody [clone: MOC-32]
Catalog # USBIS1014-57P
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Small Cell Lung Carcinoma, Neural and Endocrine Antigen
- Clonality:Monoclonal
- Clone:MOC-32
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgM
- Western blot:Yes
- Format:Supernatant
- Form:liquid
- Storage buffer:0,1% sodium azide.
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:1 ml
- Pk:1 mL
Specifications
About this item
Recommended dilution: Immunochemistry: 1:10
Type: Primary
Antigen: Small Cell Lung Carcinoma, Neural and Endocrine Antigen
Clonality: Monoclonal
Clone: MOC-32
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: