Anti-Shigella dysenteriae, Serotype 10 Mouse Monoclonal Antibody [clone: 10F429]
Catalog # USBIS1013-12D
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen symbol:Shigella dysenteriae, Serotype 10
- Clonality:Monoclonal
- Clone:10F429
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG3
- Form:liquid
- Storage buffer:PBS, 0,09% sodium azide.
- Storage temperature:Cool
- Concentration:~1 mg/ml
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:200 µg
- Pk:200 µG
Specifications
About this item
Type: Primary
Antigen: Shigella dysenteriae, Serotype 10
Clonality: Monoclonal
Clone: 10F429
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG3
Reactivity: