Anti-Serotonin Receptor 1A Mouse Monoclonal Antibody [clone: 11C254]
USBIS1001-15C
- Antibody type:Primary
- Antigen name:Serotonin Receptor 1A
- Antigen symbol:5-HT1A
- Clonality:Monoclonal
- Clone:11C254
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgM kappa
- Reactivity:Human,Rat
- Western blot:Yes
- Form:liquid
- Storage buffer:PBS, 0,05% sodium azide.
- Storage temperature:Cool
- Concentration:~1 mg/ml
- Shipping temperature:Ice
- Purification:purified by ion exchange chromatography
- Size:100 µg
- Pk:100 µG
Recommended dilution: Western Blot: 0.5 ?g/ml
Type: Primary
Antigen: Serotonin Receptor 1A
Clonality: Monoclonal
Clone: 11C254
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgM Kappa
Reactivity: Human, Rat