Anti-Secobarbital Sheep Polyclonal Antibody
USBIS0600-02A
- Antibody type:Primary
- Antigen symbol:Secobarbital
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Sheep
- Form:liquid
- Storage buffer:20mMphosphate, 150mM sodium chloride, pH 7,2, 0,09% sodium azide.
- Storage temperature:Cool
- Concentration:~~1 mg/ml (UV abs)
- Shipping temperature:Ice
- Purification:purified
- Size:100 µl
- Pk:100 µl
Recommended dilution: ELISA: 5 ?g/ml G27881
Type: Primary
Antigen: Secobarbital
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Sheep
Isotype:
Reactivity: