Anti-Salmonella sp. Mouse Monoclonal Antibody [clone: 42-77]
USBIS0060-24E
- Antibody type:Primary
- Antigen symbol:Salmonella sp.
- Clonality:Monoclonal
- Clone:42-77
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG3
- Form:liquid
- Storage buffer:PBS, 0,1% sodium azide.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by ammonium sulfate precipitation
- Size:1 ml
- Pk:1 mL
Recommended dilution: ELISA: 1:10
Type: Primary
Antigen: Salmonella sp.
Clonality: Monoclonal
Clone: 42-77
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG3
Reactivity: