Anti-SAH3 Rabbit Polyclonal Antibody
USBIS0056-57
- Antibody type:Primary
- Antigen symbol:SAH3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Form:liquid
- Storage buffer:PBS, pH 7,2, 0,09% sodium azide.
- Storage temperature:Cool
- Concentration:~5 mg/ml
- Shipping temperature:Ice
- Purification:purified by Protein G chromatography
- Size:1 ml
- Pk:1 mL
Type: Primary
Antigen: SAH3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: