Order Entry
Export
ContactUsLinkComponent
Anti-S100A9 Mouse Monoclonal Antibody [clone: 47-8D3]
Anti-S100A9 Mouse Monoclonal Antibody [clone: 47-8D3]
Catalog # USBIS0051-86A
Supplier:  US Biological
Anti-S100A9 Mouse Monoclonal Antibody [clone: 47-8D3]
Catalog # USBIS0051-86A
Supplier:  US Biological

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    S100 calcium binding protein A9
  • Antigen symbol:
    S100A9
  • Clonality:
    Monoclonal
  • Clone:
    47-8D3
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG1
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Form:
    liquid
  • Storage buffer:
    PBS, 0,05% sodium azide.
  • Storage temperature:
    Cool
  • Concentration:
    ~0,1 mg/ml
  • Shipping temperature:
    Ice
  • Purification:
    purified
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Type: Primary
Antigen: S100A9 (S100 calcium binding protein A9)
Clonality: Monoclonal
Clone: 47-8D3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human