Anti-Rhox-11 Rabbit Polyclonal Antibody
USBIR1998-90
- Antibody type:Primary
- Antigen symbol:Rhox-11
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Mouse
- Western blot:Yes
- Format:Serum
- Form:liquid
- Amino acid number:139 to 152
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µl
- Pk:100 µl
Recommended dilution: Western Blot: 1:1000-1:2000; Immunochemistry: 1:100
Type: Primary
Antigen: Rhox-11
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse