Anti-Rho Guanine Nucleotide Exchange Factor 10 Rabbit Polyclonal Antibody
Catalog # USBIR1998-28D
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Rho Guanine Nucleotide Exchange Factor 10
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human
- Form:liquid
- Storage buffer:PBS, 0,1% sodium azide.
- Storage temperature:Cool
- Concentration:~1 mg/ml
- Shipping temperature:Ice
- Size:50 µg
- Pk:50 µG
Specifications
About this item
Recommended dilution: ELISA: 1:10-1:1000; Immunochemistry: 20 ?g/ml
Type: Primary
Antigen: Rho Guanine Nucleotide Exchange Factor 10
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human