Anti-Rho Guanine Nucleotide Exchange Factor H1 Rabbit Monoclonal Antibody [clone: 7H57]
Catalog # USBIR1998-26
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Rho Guanine Nucleotide Exchange Factor H1
- Clonality:Monoclonal
- Clone:7H57
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Hamster,Monkey,Human,Rat,Mouse
- Western blot:Yes
- Format:Supernatant
- Form:liquid
- Storage buffer:10mM HEPES, pH 7,5, 150mM sodium chloride,0,1mg/ml BSA, <0,02% sodium azide, 50% glycerol .
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µl
- Pk:100 µl
Specifications
About this item
Recommended dilution: Western Blot: 1:1000; Immunoprecipitation: 1:50
Type: Primary
Antigen: Rho Guanine Nucleotide Exchange Factor H1
Clonality: monoclonal
Clone: 7H57
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Hamster, Human, Monkey, Mouse, Rat