Anti-Rabies Mouse Monoclonal Antibody [clone: RAB-50]
Catalog # USBIR0011-02
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen symbol:Rabies
- Clonality:Monoclonal
- Clone:RAB-50
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG2b
- Neutralisation:Yes
- Form:liquid
- Storage buffer:PBS, 0,05% sodium azide.
- Storage temperature:Cool
- Concentration:~0,1 mg/ml
- Shipping temperature:Ice
- Purification:purified
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Recommended dilution: ELISA: 1:50
Type: Primary
Antigen: Rabies
Clonality: monoclonal
Clone: RAB-50
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: