Anti-Protocadherin beta-15 Mouse Monoclonal Antibody [clone: 9G429]
Catalog # USBIP9119-50
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen symbol:Protocadherin beta-15
- Clonality:Monoclonal
- Clone:9G429
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Form:lyophilized powder
- Storage buffer:PBS, pH 7,2.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by affinity chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: Protocadherin beta-15
Clonality: monoclonal
Clone: 9G429
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human