Anti-PCDHA11 Mouse Polyclonal Antibody
Catalog # USBIP9119-20
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:protocadherin alpha 11
- Antigen symbol:PCDHA11
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Format:Serum
- Form:liquid
- Storage buffer:50% glycerol
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 µl
- Pk:50 µl
Specifications
About this item
Type: Primary
Antigen: PCDHA11 (protocadherin alpha 11)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human