Anti-PCDHA10 Mouse Monoclonal Antibody [clone: 9G426]
USBIP9119-19
- Antibody type:Primary
- Antigen name:Protocadherin alpha 10
- Antigen symbol:PCDHA10
- Clonality:Monoclonal
- Clone:9G426
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Form:lyophilized powder
- Storage buffer:PBS, pH 7,2
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by affinity chromatography
- Size:100 µg
- Pk:100 µG
Type: Primary
Antigen: PCDHA10 (protocadherin alpha 10)
Clonality: monoclonal
Clone: 9G426
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human