Anti-Prostaglandin J2, 15-deoxy-D12,14- Rabbit Polyclonal Antibody
Catalog # USBIP9053-35C
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen symbol:Prostaglandin J2, 15-deoxy-D12,14-
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Form:liquid
- Storage buffer:PBSwith 0,1% sodium azide.
- Storage temperature:Cool
- Concentration:1 mg/ml
- Shipping temperature:Ice
- Purification:purified by sodium sulfate precipitation
- Size:100 µl
- Pk:100 µl
Specifications
About this item
Type: Primary
Antigen: Prostaglandin J2, 15-deoxy-D12,14-
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: