Anti-Prolactin-Releasing Peptide-31 Rabbit Polyclonal Antibody
Catalog # USBIP9009-68A
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen symbol:Prolactin-Releasing Peptide-31
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat
- Form:lyophilized powder
- Sequence:SRTHRHSMEIRTPDINPAWYASRGIRPVGRF-NH2
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified
- Size:400 µg
- Pk:400 µG
Specifications
About this item
Type: Primary
Antigen: Prolactin-Releasing Peptide-31
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat