Anti-Progesterone 11 Mouse Monoclonal Antibody [clone: 8.F.235A]
Catalog # USBIP9006-10G
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen symbol:Progesterone 11
- Clonality:Monoclonal
- Clone:8.F.235A
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG2b
- Reactivity:Human
- Form:liquid
- Storage buffer:PBS, pH 7,4, 0,1% sodium azide.
- Storage temperature:Cool
- Concentration:~3 mg/ml
- Shipping temperature:Ice
- Tested applications:Radio Immunoassay
- Purification:purified by Protein A chromatography
- Size:1 mg
- Pk:1 mg
Specifications
About this item
Type: Primary
Antigen: Progesterone 11
Clonality: monoclonal
Clone: 8.F.235A
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human