Anti-Progesterone Rabbit Polyclonal Antibody
Catalog # USBIP9006-06D
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen symbol:Progesterone
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Form:liquid
- Storage buffer:PBS, 0,09% sodium azide.
- Storage temperature:Cool
- Concentration:~5 mg/ml
- Shipping temperature:Ice
- Purification:purified by Protein G chromatography
- Size:200 µl
- Pk:200 µl
Specifications
About this item
Type: Primary
Antigen: Progesterone
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: