Order Entry
Export
ContactUsLinkComponent
Anti-PR Set7 Rabbit Polyclonal Antibody
Anti-PR Set7 Rabbit Polyclonal Antibody
Catalog # USBIP5575-01B
Supplier:  US Biological
Anti-PR Set7 Rabbit Polyclonal Antibody
Catalog # USBIP5575-01B
Supplier:  US Biological

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    PR Set7
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Form:
    liquid
  • Storage buffer:
    0,1M Tris-glycine, pH 7,4, 0,15mM sodium chloride, 0,05% sodium azide.
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Purification:
    purified by immunoaffinity chromatography
  • Size:
    100 µl
  • Pk:
    100 µl

Specifications

About this item

Recommended dilution: Western Blot: 1:1000

Type: Primary
Antigen: PR Set7
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human